5-Acetyl-2-fluorphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS50124G5EA 0 CHF
ACTIAS50124G5 ACTIAS50124G1 ACTIAS50124G25
5-Acetyl-2-fluorphenylboronsäure
5-Acetyl-2-fluorphenylboronsäure
Formel: C₈H₈BFO₃
MW: 181,9567 g/mol
MDL Number: MFCD06656263
CAS-Nummer: 870777-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR