Methyl-3,5-dinitro-4-methylbenzoat

Lieferant: ACTIVATE SCIENTIFIC

AS20896G25 AS20896G5
ACTIAS20896G25EA 364 CHF
ACTIAS20896G25 ACTIAS20896G5
Methyl-3,5-dinitro-4-methylbenzoat
Methyl-3,5-dinitro-4-methylbenzoat
Formel: C₉H₈N₂O₆
MDL Number: MFCD00451040
CAS-Nummer: 49592-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR