2-(2,4-Dichlorphenoxy)benzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS24283G25 AS24283G5
ACTIAS24283G25EA 0 CHF
ACTIAS24283G25 ACTIAS24283G5
2-(2,4-Dichlorphenoxy)benzonitril
2-(2,4-Dichlorphenoxy)benzonitril
Formel: C₁₃H₇Cl₂NO
MDL Number: MFCD00052459
CAS-Nummer: 175136-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR