trans-3,3-Dimethyl-1-butenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-tert-Butyl-trans-vinylboronic acid, (E)-(3,3-Dimethylbut-1-en-1-yl)boronic acid

AS51302G1 AS51302G5
ACTIAS51302G1EA 0 CHF
ACTIAS51302G1 ACTIAS51302G5
trans-3,3-Dimethyl-1-butenylboronsäure
trans-3,3-Dimethyl-1-butenylboronic acid
Formel: C₆H₁₃BO₂
MW: 127,98 g/mol
MDL Number: MFCD01074624
CAS-Nummer: 86595-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR