1-(2-Cyanphenyl)piperidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-(1-Piperidinyl)benzonitrile, 2-Piperidinobenzonitrile

AS22083G25 AS22083G5
ACTIAS22083G25EA 281 CHF
ACTIAS22083G25 ACTIAS22083G5
1-(2-Cyanphenyl)piperidin
1-(2-Cyanphenyl)piperidin
Formel: C₁₂H₁₄N₂
MW: 186,26 g/mol
MDL Number: MFCD00049221
CAS-Nummer: 72752-52-4
EINECS: 427-330-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR