4-[4-(Trifluormethyl)phenoxy]piperidin Hydrochlorid
Lieferant: ACTIVATE SCIENTIFIC
AS30181G1
AS30181G5
ACTIAS30181G1EA
0
CHF
ACTIAS30181G1
ACTIAS30181G5
4-[4-(Trifluormethyl)phenoxy]piperidin Hydrochlorid
4-[4-(Trifluormethyl)phenoxy]piperidin Hydrochlorid
Formel:
C₁₂H₁₅ClF₃NO₂ |
MDL Number:
MFCD02258937 CAS-Nummer: 287952-09-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...