5-(Tributylstannyl)thiazol

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

AS50126G1 AS50126G5
ACTIAS50126G1EA 183 CHF
ACTIAS50126G1 ACTIAS50126G5
5-(Tributylstannyl)thiazol
5-(Tributylstannyl)thiazol
Formel: C₁₅H₂₉NSSn
MDL Number: MFCD01319074
CAS-Nummer: 157025-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR