5-Methyl-2-(tributylstannyl)thiazol

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

AS51234G1 AS51234G5
ACTIAS51234G1EA 393 CHF
ACTIAS51234G1 ACTIAS51234G5
5-Methyl-2-(tributylstannyl)thiazol
5-Methyl-2-(tributylstannyl)thiazol
Formel: C₁₆H₃₁NSSn
MW: 388,19 g/mol
MDL Number: MFCD09025809
CAS-Nummer: 848613-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR