2-(N-Boc)-5-(tributylstannyl)thiazol

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

AS50133G1 AS50133G5
ACTIAS50133G1EA 0 CHF
ACTIAS50133G1 ACTIAS50133G5
2-(N-Boc)-5-(tributylstannyl)thiazol
2-(N-Boc)-5-(tributylstannyl)thiazol
Formel: C₂₀H₃₈N₂O₂SSn
MDL Number: MFCD07787377
CAS-Nummer: 243972-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR