2-(Methylthio)-4-(tributylstannyl)pyrimidin

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

AS50128G1 AS50128G5
ACTIAS50128G1EA 0 CHF
ACTIAS50128G1 ACTIAS50128G5
2-(Methylthio)-4-(tributylstannyl)pyrimidin
2-(Methylthio)-4-(tributylstannyl)pyrimidin
Formel: C₁₇H₃₂N₂SSn
MDL Number: MFCD01319026
CAS-Nummer: 123061-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR