2-Ethoxy-5-(tributylstannyl)pyrimidin

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

AS51232G1 AS51232G5
ACTIAS51232G1EA 297 CHF
ACTIAS51232G1 ACTIAS51232G5
2-Ethoxy-5-(tributylstannyl)pyrimidin
2-Ethoxy-5-(tributylstannyl)pyrimidin
Formel: C₁₈H₃₄N₂OSn
MDL Number: MFCD09025789
CAS-Nummer: 1025746-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR