2-Chlor-5-(tributylstannyl)pyrimidin

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

AS51173G1 AS51173G5
ACTIAS51173G1EA 227 CHF
ACTIAS51173G1 ACTIAS51173G5
2-Chlor-5-(tributylstannyl)pyrimidin
2-Chlor-5-(tributylstannyl)pyrimidin
Formel: C₁₆H₂₉ClN₂Sn
MDL Number: MFCD07787378
CAS-Nummer: 155191-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR