2-(Tributylstannyl)pyrazin
Lieferant: ACTIVATE SCIENTIFIC
|
Gefahr
|
AS50213G1
AS50213G5
ACTIAS50213G1EA
147
CHF
ACTIAS50213G1
ACTIAS50213G5
2-(Tributylstannyl)pyrazin
2-(Tributylstannyl)pyrazin
Formel:
C₁₆H₃₀N₂Sn MW: 369.12 g/mol |
MDL Number:
MFCD01319077 CAS-Nummer: 205371-27-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...