2-(Tributylstannyl)oxazol
Lieferant: ACTIVATE SCIENTIFIC
|
Gefahr
|
AS51611G1
AS51611G5
ACTIAS51611G1EA
162
CHF
ACTIAS51611G1
ACTIAS51611G5
2-(Tributylstannyl)oxazol
2-(Tributylstannyl)oxazol
Formel:
C₁₅H₂₉NOSn MW: 358,11 g/mol |
MDL Number:
MFCD06798076 CAS-Nummer: 145214-05-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...