Cholinchlorid-[Trimethyl-D9] (98% D)

Lieferant: CAMBRIDGE ISOTOPE LABS., INC.

CAMBDLM-549-1EA 0 CHF
CAMBDLM-549-1
Cholinchlorid-[Trimethyl-D9] (98% D)
Cholinchlorid-[Trimethyl-D9]
Formel: C₅H₅D₉ClNO
MDL Number: MFCD00143968
CAS-Nummer: 61037-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR