Methyl-6-chlor-3-methylpicolinat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 6-chloro-3-methylpicolinate, Methyl 6-chloro-3-methylpyridine-2-carboxylate

AS3261G1 AS3261G5
ACTIAS3261G1EA 0 CHF
ACTIAS3261G1 ACTIAS3261G5
Methyl-6-chlor-3-methylpicolinat
Methyl-6-chlor-3-methylpicolinat
Formel: C₈H₈ClNO₂
MW: 185,6076 g/mol
MDL Number: MFCD12756112
CAS-Nummer: 878207-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR