Ethyl-4-chlorchinolin-3-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Chloroquinoline-3-carboxylic acid ethyl ester, Ethyl 4-chloroquinoline-3-carboxylate

AS20755G1 AS20755G5
ACTIAS20755G1EA 43 CHF
ACTIAS20755G1 ACTIAS20755G5
Ethyl-4-chlorchinolin-3-carboxylat
Ethyl-4-chlorchinolin-3-carboxylat
Formel: C₁₂H₁₀ClNO₂
MDL Number: MFCD00173407
CAS-Nummer: 13720-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR