5-(Methoxycarbonyl)-2-methylphenylboronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 4-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

AS16225G1 AS16225G5
ACTIAS16225G1EA 0 CHF
ACTIAS16225G1 ACTIAS16225G5
5-(Methoxycarbonyl)-2-methylphenylboronsäurepinakolester
5-(Methoxycarbonyl)-2-methylphenylboronsäurepinakolester
Formel: C₁₅H₂₁BO₄
MW: 276,136 g/mol
MDL Number: MFCD11520530
CAS-Nummer: 882679-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR