5-Methoxy-3-pyridinboronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Methoxy-3-pyridylboronic acid Pinacol ester, 3-Methoxypyridine-5-boronic acid Pinacol ester, 3-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(5-Methoxy-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

AS50875G1 AS50875G5
ACTIAS50875G1EA 0 CHF
ACTIAS50875G1 ACTIAS50875G5
5-Methoxy-3-pyridinboronsäurepinakolester
5-Methoxy-3-pyridinboronsäurepinakolester
Formel: C₁₂H₁₈BNO₃
MW: 235,0872 g/mol
MDL Number: MFCD03412068
CAS-Nummer: 445264-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR