3-Amino-2-chlor-5-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS31822G1 AS31822G5
ACTIAS31822G1EA 0 CHF
ACTIAS31822G1 ACTIAS31822G5
3-Amino-2-chlor-5-methylpyridin
3-Amino-2-chloro-5-methylpyridine
Formel: C₆H₇ClN₂
MDL Number: MFCD03427656
CAS-Nummer: 34552-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR