6-Amino-5-chlornicotinonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Amino-3-chloro-5-cyanopyridine

AS30386G1 AS30386G5
ACTIAS30386G1EA 0 CHF
ACTIAS30386G1 ACTIAS30386G5
6-Amino-5-chlornicotinonitril
6-Amino-5-chlornicotinonitril
Formel: C₆H₄ClN₃
MW: 153,5691 g/mol
MDL Number: MFCD03617768
CAS-Nummer: 156361-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR