5-Cyan-2-fluorbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Fluoro-3-formylbenzonitrile

AS33489G1 AS33489G10 AS33489G5
ACTIAS33489G1EA 0 CHF
ACTIAS33489G1 ACTIAS33489G10 ACTIAS33489G5
5-Cyan-2-fluorbenzaldehyd
5-Cyan-2-fluorbenzaldehyd
Formel: C₈H₄FNO
MW: 149,1219 g/mol
MDL Number: MFCD04974126
CAS-Nummer: 146137-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR