Methyl-2-(brommethyl)benzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 2-bromomethylbenzoate

AS32325G1 AS32325G5
ACTIAS32325G1EA 0 CHF
ACTIAS32325G1 ACTIAS32325G5
Methyl-2-(brommethyl)benzoat
Methyl-2-(brommethyl)benzoat
Formel: C₉H₉BrO₂
MW: 229,0706 g/mol
MDL Number: MFCD03425900
CAS-Nummer: 2417-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR