Methyl-2-(brommethyl)-4-methoxybenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 2-(bromomethyl)-4-methoxybenzoate

AS31178G1 AS31178G5
ACTIAS31178G1EA 0 CHF
ACTIAS31178G1 ACTIAS31178G5
Methyl-2-(brommethyl)-4-methoxybenzoat
Methyl-2-(brommethyl)-4-methoxybenzoat
Formel: C₁₀H₁₁BrO₃
MW: 259.1000 g/mol
MDL Number: MFCD04114316
CAS-Nummer: 15365-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR