Methyl-2-(brommethyl)-3-chlorbenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 2-(bromomethyl)-3-chlorobenzoate

AS34368G1 AS34368G5
ACTIAS34368G1EA 0 CHF
ACTIAS34368G1 ACTIAS34368G5
Methyl-2-(brommethyl)-3-chlorbenzoat
Methyl-2-(brommethyl)-3-chlorbenzoat
Formel: C₉H₈BrClO₂
MW: 263.5156 g/mol
MDL Number: MFCD04114322
CAS-Nummer: 188187-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR