Isochinolin-8-amin

Lieferant: ACTIVATE SCIENTIFIC

AS1282G1 AS1282G5
ACTIAS1282G1EA 0 CHF
ACTIAS1282G1 ACTIAS1282G5
Isochinolin-8-amin
Isochinolin-8-amin
Formel: C₉H₈N₂
MDL Number: MFCD00179553
CAS-Nummer: 23687-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR