Isochinolin-6-amin

Lieferant: ACTIVATE SCIENTIFIC

AS1280G1 AS1280G5
ACTIAS1280G1EA 0 CHF
ACTIAS1280G1 ACTIAS1280G5
Isochinolin-6-amin
Isochinolin-6-amin
Formel: C₉H₈N₂
MDL Number: MFCD04114862
CAS-Nummer: 23687-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR