7-Isochinolinol

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 7-Hydroxyisoquinoline, Isoquinolin-7-ol

AS1284G1 AS1284G5
ACTIAS1284G1EA 0 CHF
ACTIAS1284G1 ACTIAS1284G5
7-Isochinolinol
7-Isochinolinol
Formel: C₉H₇NO
MW: 145,16 g/mol
MDL Number: MFCD00456131
CAS-Nummer: 7651-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR