6-Isochinolinol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Hydroxyisoquinoline, Isoquinolin-6-ol

AS1283G1 AS1283G5
ACTIAS1283G1EA 0 CHF
ACTIAS1283G1 ACTIAS1283G5
6-Isochinolinol
6-Isochinolinol
Formel: C₉H₇NO
MW: 145,16 g/mol
MDL Number: MFCD04114860
CAS-Nummer: 7651-82-3
EINECS: 231-612-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR