5-Isochinolinol

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 5-Hydroxyisoquinoline

AS30967G1 AS30967G5
ACTIAS30967G1EA 0 CHF
ACTIAS30967G1 ACTIAS30967G5
5-Isochinolinol
5-Isochinolinol
Formel: C₉H₇NO
MW: 145.16 g/mol
MDL Number: MFCD00006906
CAS-Nummer: 2439-04-5
EINECS: 219-456-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR