5-Brom-3-pyridinboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Bromopyridine-5-boronic acid, 3-Bromopyridyl-5-boronic acid

AS50231G1 AS50231G5
ACTIAS50231G1EA 0 CHF
ACTIAS50231G1 ACTIAS50231G5
5-Brom-3-pyridinboronsäure
5-Bromo-3-pyridineboronic acid
Formel: C₅H₅BBrNO₂
MW: 201.81 g/mol
MDL Number: MFCD02685634
CAS-Nummer: 452972-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR