4-Brom-3-iodbenzotrifluorid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Bromo-3-iodobenzotrifluoride, 1-Bromo-2-iodo-4-(trifluoromethyl)benzene

AS34898G1 AS34898G5
ACTIAS34898G1EA 0 CHF
ACTIAS34898G1 ACTIAS34898G5
4-Brom-3-iodbenzotrifluorid
4-Brom-3-iodbenzotrifluorid
Formel: C₇H₃BrF₃I
MW: 350,9 g/mol
MDL Number: MFCD09800691
CAS-Nummer: 640280-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR