Natriumdodecylsulfat (SDS) 0,50%, DILUT-IT® Auflösungsmedium-Konzentrat, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
Synonyms:
Dodecylschwefelsäure Natriumsalz
, Natriumlaurylsulfat
, Dodecylnatriumsulfat
, Schwefelsäuredodecylester Natriumsalz
, Laurylsulfat Natriumsalt
Veränderungsmanagement-Kategorie = N
D018-01EA
47
CHF
D018-01
Natriumdodecylsulfat (SDS) 0,50%, DILUT-IT® Auflösungsmedium-Konzentrat, J.T.Baker®
Natriumdodecylsulfat (SDS)
DILUT-IT dissolution media concentrates are produced from raw materials in accordance with USP/EP/BP/JP guidelines under validated processes in Avantor Performance Materials' ISO certified manufacturing facilities
Formel:
CH₃(CH₂)₁₁OSO₃Na MW: 288,38 g/mol Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00036175 CAS-Nummer: 151-21-3 EINECS: 205-788-1 |
![]() |
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use | |
STORE AT ROOM TEMPERATURE | |
Appearance | Passes Test |
Assay (at 15x Dilution) | 0.49 - 0.51 % |
Density (g/mL) at 25°C, For Information Only | |
DILUT-IT dissolution media concentrates are produced from raw materials in accordance with USP/EP/BP/JP guidelines under validated processes in Avantor Performance Materials' ISO certified manufacturing facilities |
Learn more
![](/m-ch.vwr.com/de_CH/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...