Ethyl-3-iodbenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Iodobenzoic acid ethyl ester

AS34876G10 AS34876G100
ACTIAS34876G10EA 0 CHF
ACTIAS34876G10 ACTIAS34876G100
Ethyl-3-iodbenzoat
Ethyl-3-iodbenzoat
Formel: C₉H₉IO₂
MW: 276.07 g/mol
Siedepunkt: 165
Dichte: 1.66 g/cm³ (20 °C)
MDL Number: MFCD00044769
CAS-Nummer: 58313-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR