6-Fluorsalicylsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Fluoro-2-hydroxybenzoic acid, 2-Fluoro-6-hydroxybenzoic acid

AS35082G1 AS35082G10 AS35082G5
ACTIAS35082G1EA 0 CHF
ACTIAS35082G1 ACTIAS35082G10 ACTIAS35082G5
6-Fluorsalicylsäure
6-Fluorsalicylsäure
Formel: C₇H₅FO₃
MW: 156,11 g/mol
MDL Number: MFCD00052179
CAS-Nummer: 67531-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR