(3-Methoxyphenyl)essigsäure

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

Synonyms: 3-Methoxyphenylacetic acid

AS20083G100 AS20083G25
ACTIAS20083G100EA 127 CHF
ACTIAS20083G100 ACTIAS20083G25
(3-Methoxyphenyl)essigsäure
(3-Methoxyphenyl)essigsäure
Formel: (C₆H₄(OCH₃))CH₂COOH
MW: 166,18 g/mol
Siedepunkt: 305.95 °C (1013 hPa)
Melting Pt: 71…73 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00004334
CAS-Nummer: 1798-09-0
EINECS: 217-282-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR