(2,6-Difluor-4-methoxyphenyl)essigsäure
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
AS33449G1
AS33449G5
ACTIAS33449G1EA
0
CHF
ACTIAS33449G1
ACTIAS33449G5
(2,6-Difluor-4-methoxyphenyl)essigsäure
(2,6-Difluor-4-methoxyphenyl)essigsäure
Formel:
(C₆H₂F₂OCH₃)CH₂COOH MW: 186,16 g/mol |
MDL Number:
MFCD04115927 CAS-Nummer: 886498-98-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...