4-Methyl-3-pyridinboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (4-Methylpyridin-3-yl)boronic acid, 4-Methylpyridine-3-boronic acid

AS50703G1 AS50703G5
ACTIAS50703G1EA 0 CHF
ACTIAS50703G1 ACTIAS50703G5
4-Methyl-3-pyridinboronsäure
4-Methyl-3-pyridineboronic acid
Formel: C₆H₈BNO₂
MW: 136.94 g/mol
MDL Number: MFCD03788244
CAS-Nummer: 148546-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR