5-Methoxypyridin-3-boronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-methoxypyridin-3-yl boronic acid, (5-Methoxypyridin-3-yl)boronic acid, 5-Methoxy-3-pyridineboronic acid

AS33593G1 AS33593G5
ACTIAS33593G1EA 0 CHF
ACTIAS33593G1 ACTIAS33593G5
5-Methoxypyridin-3-boronsäure
5-Methoxypyridin-3-boronsäure
Formel: C₆H₈BNO₃
MW: 152.95 g/mol
MDL Number: MFCD03425977
CAS-Nummer: 850991-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR