Methyl-2-(brommethyl)-4-chlorbenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 2-(bromomethyl)-4-chlorobenzoate

AS34367G1 AS34367G5
ACTIAS34367G1EA 0 CHF
ACTIAS34367G1 ACTIAS34367G5
Methyl-2-(brommethyl)-4-chlorbenzoat
Methyl-2-(brommethyl)-4-chlorbenzoat
Formel: C₉H₈BrClO₂
MW: 263.5156 g/mol
MDL Number: MFCD11505950
CAS-Nummer: 145908-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR