2,4-Dichlor-6-fluorchinolin

Lieferant: ACTIVATE SCIENTIFIC

AS36773G1 AS36773G5
ACTIAS36773G1EA 0 CHF
ACTIAS36773G1 ACTIAS36773G5
2,4-Dichlor-6-fluorchinolin
2,4-Dichloro-6-fluoroquinoline
Formel: C₉H₄Cl₂FN
MDL Number: MFCD09261383
CAS-Nummer: 406204-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR