Methyl-4-amino-2-methylbenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 4-amino-2-methylbenzoate

AS1481G1 AS1481G5
ACTIAS1481G1EA 0 CHF
ACTIAS1481G1 ACTIAS1481G5
Methyl-4-amino-2-methylbenzoat
Methyl-4-amino-2-methylbenzoat
Formel: C₉H₁₁NO₂
MDL Number: MFCD08458829
CAS-Nummer: 6933-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR