Methyl-3-amino-4-methylbenzoat
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
Methyl 3-amino-4-methylbenzoate
AS20944G100
AS20944G25
ACTIAS20944G100EA
0
CHF
ACTIAS20944G100
ACTIAS20944G25
Methyl-3-amino-4-methylbenzoat
Methyl-3-amino-4-methylbenzoat
Formel:
C₉H₁₁NO₂ MW: 165.19 g/mol |
MDL Number:
MFCD00025206 CAS-Nummer: 18595-18-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...