Methyl-3-amino-2-methylbenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 3-amino-2-methylbenzoate, 3-Amino-o-toluic acid Methyl Ester, 3-Amino-2-methylbenzoic acid Methyl Ester

AS24286G100 AS24286G25
ACTIAS24286G100EA 0 CHF
ACTIAS24286G100 ACTIAS24286G25
Methyl-3-amino-2-methylbenzoat
Methyl 3-amino-2-methylbenzoate
Formel: C₉H₁₁NO₂
MW: 165,19 g/mol
MDL Number: MFCD00211328
CAS-Nummer: 18583-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR