Methyl-3-amino-4-iodbenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 3-amino-4-iodobenzoate

AS35048G1 AS35048G10 AS35048G5
ACTIAS35048G1EA 0 CHF
ACTIAS35048G1 ACTIAS35048G10 ACTIAS35048G5
Methyl-3-amino-4-iodbenzoat
Methyl-3-amino-4-iodbenzoat
Formel: C₈H₈INO₂
MW: 277,06 g/mol
MDL Number: MFCD08064225
CAS-Nummer: 412947-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR