Ethyl-4-amino-3-fluorbenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl 4-amino-3-fluorobenzoate

AS33367G1 AS33367G10 AS33367G5
ACTIAS33367G1EA 0 CHF
ACTIAS33367G1 ACTIAS33367G10 ACTIAS33367G5
Ethyl-4-amino-3-fluorbenzoat
Ethyl-4-amino-3-fluorbenzoat
Formel: C₉H₁₀FNO₂
MW: 183,1796 g/mol
MDL Number: MFCD07782043
CAS-Nummer: 73792-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR