Methyl-4-amino-2-fluorbenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 4-amino-2-fluorobenzoate

AS34897G1 AS34897G5
ACTIAS34897G1EA 0 CHF
ACTIAS34897G1 ACTIAS34897G5
Methyl-4-amino-2-fluorbenzoat
Methyl-4-amino-2-fluorbenzoat
Formel: C₈H₈FNO₂
MW: 169,1530 g/mol
MDL Number: MFCD09037917
CAS-Nummer: 73792-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR