Methyl-2-amino-4-fluorbenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 2-amino-4-fluorobenzoate

AS22320G1 AS22320G25 AS22320G5
ACTIAS22320G1EA 21 CHF
ACTIAS22320G1 ACTIAS22320G25 ACTIAS22320G5
Methyl-2-amino-4-fluorbenzoat
Methyl-2-amino-4-fluorbenzoat
Formel: C₈H₈FNO₂
MW: 169,15 g/mol
MDL Number: MFCD11054082
CAS-Nummer: 2475-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR