Methyl-2-amino-3-fluorbenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 2-amino-3-fluorobenzoate

AS34854G1 AS34854G10 AS34854G5
ACTIAS34854G1EA 0 CHF
ACTIAS34854G1 ACTIAS34854G10 ACTIAS34854G5
Methyl-2-amino-3-fluorbenzoat
Methyl-2-amino-3-fluorbenzoat
Formel: C₈H₈FNO₂
MW: 169,1530 g/mol
MDL Number: MFCD11870179
CAS-Nummer: 144851-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR