Ethyl-2-amino-4,5-dichlorbenzoat

Lieferant: ACTIVATE SCIENTIFIC

AS34625G1 AS34625G5
ACTIAS34625G1EA 0 CHF
ACTIAS34625G1 ACTIAS34625G5
Ethyl-2-amino-4,5-dichlorbenzoat
Ethyl-2-amino-4,5-dichlorbenzoat
Formel: C₉H₉Cl₂NO₂
MDL Number: MFCD10566266
CAS-Nummer: 1108668-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR